missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LECT2 Antibody (MM0449-4B24) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11768-0.025MG
This item is not returnable.
View return policy
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LECT2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100-1:2000, Immunohistochemistry | |
| O14960 | |
| LECT2 | |
| Human LECT2 | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| MM0449-4B24 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3950 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction