missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Leadmium™ Green AM dye
Click to view available options
Quantity:
5 x 50 μg
Unit Size:
250µg
Description
The proprietary Leadmium™ Green AM dye from Molecular Probes™ provides researchers with a unique tool to specifically measure intracellular lead and cadmium levels with high sensitivity. While standards assays exist for measuring lead and cadmium in solution, the unique Leadmium™ Green AM indicator was specifically designed to detect these toxic environmental contaminants inside living cells.
–Highly specific indicator for intracellular measurements of lead and cadmium
–Flexible format allows use on multiple formats - flow cytometry, fluorescence microscopy imaging and plate readers
–Bright fluorescence and high discrimination from calcium allows detection of nanomolar levels of lead and micromolar levels of cadmium
–Convenient excitation with 488 nm laser line
–The Leadmium™ Green AM dye is readily loaded and is retained very well following esterase cleavage of the acetoxymethyl group
–Highly specific indicator for intracellular measurements of lead and cadmium
–Flexible format allows use on multiple formats - flow cytometry, fluorescence microscopy imaging and plate readers
–Bright fluorescence and high discrimination from calcium allows detection of nanomolar levels of lead and micromolar levels of cadmium
–Convenient excitation with 488 nm laser line
–The Leadmium™ Green AM dye is readily loaded and is retained very well following esterase cleavage of the acetoxymethyl group
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Dye Type | Fluorescent Dye-Based |
| Product Line | Leadmium |
| Quantity | 5 x 50 μg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction