missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LDS 751

Product Code. 11544986
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11544986

Brand: Molecular Probes™ L7595

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LDS 751

LDS 751 is a cell-permeant nucleic acid stain that has a peak excitation at ∼543 nm on dsDNA and is particular useful in multicolor analyses due to its long wavelength emission maximum (∼712 nm). Binding of LDS 751 to dsDNA results in a ∼20-fold fluorescence enhancement.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 vial of LDS 751 (10 mg). Store at room temperature and protect from light.
Excitation Wavelength Range 543⁄712
Dye Type LDS 751
For Use With (Equipment) Flow Cytometer
Quantity 10 mg
Detection Method Fluorescence
Form Solid
Shipping Condition Room Temperature
Product Type Nucleic Acid Stain
SubCellular Localization Nucleic Acids, Nucleus

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.