missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDHAL6B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55494
This item is not returnable.
View return policy
Description
LDHAL6B Polyclonal specifically detects LDHAL6B in Human samples. It is validated for Western Blot.
Specifications
| LDHAL6B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 1.1.1, lactate dehydrogenase A-like 6, lactate dehydrogenase A-like 6B, LDH6B, LDHAL6, LDHLEC 1.1.1.27, L-lactate dehydrogenase A-like 6B | |
| Rabbit | |
| 42 kDa | |
| 100 μL | |
| Primary | |
| Bovine 79%, Porcine 79%. | |
| Human, Rat, Pig, Bovine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BYZ2 | |
| LDHAL6B | |
| Synthetic peptides corresponding to LDHAL6B(lactate dehydrogenase A-like 6B) The peptide sequence was selected from the middle region of LDHAL6B. Peptide sequence SGVNIAGVPLKDLNSDIGTDKDPEQWKNVHKEVTATAYEIIKMKGYTSWA. | |
| Affinity purified | |
| RUO | |
| 92483 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction