missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDHAL6B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | LDHAL6B |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232071
|
Novus Biologicals
NBP3-35488-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233044
|
Novus Biologicals
NBP3-35488-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LDHAL6B Polyclonal antibody specifically detects LDHAL6B in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| LDHAL6B | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| EC 1.1.1, lactate dehydrogenase A-like 6, lactate dehydrogenase A-like 6B, LDH6B, LDHAL6, LDHLEC 1.1.1.27, L-lactate dehydrogenase A-like 6B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human LDHAL6B (NP_149972.1).,, Sequence:, MSWTVPVVRASQRVSSVGANFLCLGMALCPRQATRIPLNGTWLFTPVSKMATVKSELIERFTSEKPVHHSKVSIIGTGSV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 92483 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title