missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54998
This item is not returnable.
View return policy
Description
LDB3 Polyclonal specifically detects LDB3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| LDB3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CMD1C, CYPHER, KIAA01613, KIAA0613FLJ35865, LDB3Z1, LDB3Z4, LIM domain binding 3, LIM domain-binding protein 3, ORACLE, PDLIM6, PDZ and LIM domain 6, Protein cypher, ZASPLVNC3, Z-band alternatively spliced PDZ-motif protein | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O75112-5 | |
| LDB3 | |
| Synthetic peptides corresponding to LDB3(LIM domain binding 3) The peptide sequence was selected from the N terminal of LDB3. Peptide sequence PVIPHQKDPALDTNGSLVAPSPSPEARASPGTPGTPELRPTFSPAFSRPS. | |
| Protein A purified | |
| RUO | |
| 11155 | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction