missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | LC3C |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282651
|
Novus Biologicals
NBP2-57604 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677277
|
Novus Biologicals
NBP2-57604-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LC3C Polyclonal specifically detects LC3C in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| LC3C | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Cellular Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 440738 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TKFLVPQELTMTQFLSIIRSRMVLRATEAF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| ATG8J, Autophagy-related protein LC3 C, Autophagy-related ubiquitin-like modifier LC3 C, LC3C, LC3-like protein 2, MAP1 light chain 3-like protein 2, MAP1 light chain 3-like protein 3, MAP1A/MAP1B LC3 C, microtubule-associated protein 1 light chain 3 gammaMAP1A/MAP1B light chain 3 C, microtubule-associated proteins 1A/1B light chain 3C | |
| MAP1LC3C | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title