missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ TLC Spray Visualization Reagents

Product Code. 10650152 Shop All Macherey-Nagel Products
Click to view available options
Chemical Name or Material:
2'7'-Dichlorofluorescein
4-(Dimethylamino)-benzaldehyde
Aniline phthalate
Bromocresol green
Caffeine detection reagent
Dragendorff-Munier
Iron(III) Chloride
Molydatophosphoric acid
Ninhydrin
Potassium Ferricyanide
Rhodamin B
Rubeanic acid
Unit Size:
1 set
100mL
This item is not returnable. View return policy

Product Code. 10650152

Brand: Macherey-Nagel™ 814404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.

  • Broad selection of premium reagents available for detection of a wide range of substances
  • Can be used for spraying or dipping of TLC plates
  • Packaged in 100mL screw-top amber bottles
  • Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)

Specifications

Chemical Name or Material Potassium Ferricyanide
CAS 7705-08-0
Color Slightly yellow
Detection Method Spraying or Dipping
Concentration 1 to <10% Potassium Hexacyanoferrate(III)
Formulation Potassium Hexacyanoferrate(III) in Water
For Use With (Application) Detection of acetylsalicylic acid, paracetamol
For Use With (Equipment) TLC plates
Matrix Water
Molecular Formula C6FeK3N6
Odor Odorless
Packaging Type Amber bottle with screw cap
Product Type Spray Visualization Reagent
Quantity 100 mL
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.