missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lauryl Tryptose Broth with MUG and Tryptophan (with Durham Tube)

Product Code. 15341302
Change view
Click to view available options
Fill Weight:
9.5 to 10.5 g
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Unit Size
15341302 9.5 to 10.5 g 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15341302 Supplier Thermo Scientific™ Supplier No. TV5079E
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate presumptive Escherichia coli as well as other coliforms, using the most portable number (MPN) method using this chromogenic media.

Enumerate presumptive Escherichia coli as well as other coliforms, using the most portable number (MPN) method using Lauryl Tryptose Broth with MUG and Tryptophan (with Durham Tube).

TRUSTED_SUSTAINABILITY

Specifications

Color Maize Yellow, Transparent
Content And Storage 2°C to 12°C
Format Poured Tube
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Weight 9.5 to 10.5 g
Packaging Type 50 Tubes/Box
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.