missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Antibody (SR1593), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | LAT |
|---|---|
| Clone | SR1593 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621384
|
Novus Biologicals
NBP3-22544-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603704
|
Novus Biologicals
NBP3-22544-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| LAT | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| A synthesized peptide derived from human LAT (Uniprot #: O43561) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1593 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 27040 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title