missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Antibody (SR1593), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22544-100ul
This item is not returnable.
View return policy
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| LAT | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1593 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| A synthesized peptide derived from human LAT (Uniprot #: O43561) | |
| 100 μg | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 27040 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction