missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | LASS4 |
|---|---|
| Dilution | Western Blot : 1:400 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066787
|
Novus Biologicals™
NBP3-30904-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067131
|
Novus Biologicals™
NBP3-30904-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LASS4 Polyclonal antibody specifically detects LASS4 in Human samples. It is validated for Western BlotSpecifications
| LASS4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CerS4, FLJ12089, LAG1 homolog, ceramide synthase 4, LAG1 longevity assurance homolog 4, LAG1 longevity assurance homolog 4 (S. cerevisiae), Trh1 | |
| Rabbits were immunized with a synthetic peptide derived from human LASS4 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 79603 | |
| IgG | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title