missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30904-25ug
This item is not returnable.
View return policy
Description
LASS4 Polyclonal antibody specifically detects LASS4 in Human samples. It is validated for Western Blot
Specifications
| LASS4 | |
| Polyclonal | |
| Western Blot : 1:400 | |
| CerS4, FLJ12089, LAG1 homolog, ceramide synthase 4, LAG1 longevity assurance homolog 4, LAG1 longevity assurance homolog 4 (S. cerevisiae), Trh1 | |
| Rabbits were immunized with a synthetic peptide derived from human LASS4 protein. | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 79603 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction