Learn More
Gilac™ Large Volume Semicircular Container
Product Code. 17938521
Change view
Click to view available options
Capacity (Metric):
170 L
220 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
17938521 220 L Each
17998411 170 L Each
2 options
Product Code. 17938521
Brand: Gilac™ G145321
Only null left
Add to basket
Add to basket
Large Volume Semicircular Container is ideal for culinary preparations. Extremely durable: tough and very strong.
  • Large volume semicircular container.
  • Ideal for culinary preparations.
  • Extremely durable.
  • Resistance to chemical products.
  • Tough and very strong.
  • Compatible with big dough container trolleys.
  • Standing work height.
  • Volume in litres is equivalent to the container filled to the brim.
Specifications
Type Storage Tote
Material HDPE
Color White
Shape Semi-circular
Capacity (English) 58.1 gal.
Capacity (Metric) 220 L