missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAR/PTPRF Antibody (S165-38), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £470.00
Specifications
| Antigen | LAR/PTPRF |
|---|---|
| Clone | S165-38 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690928
|
Novus Biologicals
NBP2-42172 |
0.1 mg |
£470.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646857
|
Novus Biologicals
NBP2-42172-0.025mg |
0.025 mg |
£176.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAR/PTPRF Monoclonal antibody specifically detects LAR/PTPRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| LAR/PTPRF | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Signaling, Neuroscience | |
| PBS (pH 7.4), 50% Glycerol | |
| LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F | |
| Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR. 97% identical in both rat and mouse. >80% identity with PTPRD and PTPRS. >50% identity with PTPRM and PTPRK. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S165-38 | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P10586 | |
| 5792 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title