missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAR/PTPRF Antibody (S165-38), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42172
This item is not returnable.
View return policy
Description
LAR/PTPRF Monoclonal antibody specifically detects LAR/PTPRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| LAR/PTPRF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence | |
| P10586 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| S165-38 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F | |
| Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR. 97% identical in both rat and mouse. >80% identity with PTPRD and PTPRS. >50% identity with PTPRM and PTPRK. | |
| 0.1 mg | |
| Cellular Signaling, Neuroscience | |
| 5792 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction