missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAPTM4B Antibody (CL5287), Novus Biologicals™
Mouse Monoclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | LAPTM4B |
|---|---|
| Clone | CL5287 |
| Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254074
|
Novus Biologicals
NBP2-61157 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613578
|
Novus Biologicals
NBP2-61157-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAPTM4B Monoclonal specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LAPTM4B | |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human | |
| 55353 | |
| This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:DTTVLLPPYDDATVNGAAKE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL5287 | |
| Monoclonal | |
| Purified | |
| Stem Cell Markers | |
| LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta | |
| LAPTM4B | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title