missing translation for 'onlineSavingsMsg'
Learn More

LAPTM4B Antibody (CL5287), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61157-25ul

 View more versions of this product

Product Code. 18613578

  • £328.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LAPTM4B Monoclonal specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAPTM4B
Monoclonal
Unconjugated
PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide
LAPTM4B
This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:DTTVLLPPYDDATVNGAAKE
25 μL
Stem Cell Markers
55353
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL5287
Immunohistochemistry-Paraffin 1:200 - 1:500
LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta
Mouse
Protein A purified
RUO
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.