missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-pSer (PKC) Antibody Kit

A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.

Brand:  Invitrogen™ PV3560

 View more versions of this product

Product Code. 11563457

  • £3940.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.

For use in combination with the Fluorescein-PKC substrate (Cat. No. PV3506) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serine, phospho (PKC Substrate)
0.96mg/mL
150kDa
RUO
Primary
Terbium-labeled anti-pSer (PKC Substrate) antibody in HEPES buffered saline.

Store at -20°C.
Biochemical terbium phospho-specific activity
340/490, 520 nm
Dry Ice
Hu
Serine/Threonine Kinases
AURKB, DYRK4, MARK4, MAP4K2, MAP4K4, MAP4K5, MINK1, MST4, NEK2, PHKG1, PKC, PRKX, ROCK2, RPS6KA1, RPS6KA2, STK24, STK25, SGK, SGK2, SGKL, TAOK2, TBK1
Kinase Assay
Tb (Terbium)/FITC
25 μg
Protein Assays and Analysis
1 Kit(s)
150,000 Da
384-well Plate
Human
End Point
MAP4K5, PRKCH, PRKCQ, PRKX, ROCK2, PRKCD, NEK2, PRKCG, SGK, MAP4K2, MINK1, SGK2, PRKCA, PRKCI, AURKB, TAOK2, MAP4K4, MARK4, DYRK4, PHKG1, PRKCZ, ROCK1, RPS6KA1, PRKCB1, STK24, PRKCB2, RPS6KA2, SGKL, STK25, PRKCE, MST4
TR-FRET
LanthaScreen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.