Learn More
Invitrogen™ LanthaScreen™ Tb-pSer (PKC) Antibody Kit
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
£3940.00 - £19140.00
Specifications
Reactivity Human
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11563457
View Documents
Invitrogen™
PV3560
25 μg
£3940.00
25µg
View alternative products
Save Up to 
»
10514013
View Documents
Invitrogen™
PV3561
1 mg
£19140.00
1mg
View alternative products
Save Up to 
»
Description
Description
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
For use in combination with the Fluorescein-PKC substrate (Cat. No. PV3506) to evaluate inhibitors in medium to high throughput screening applications.