missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody Kit
For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications.
£3530.00 - £14220.00
Specifications
| Conjugate | Tb (Terbium)/GFP (EmGFP) |
|---|---|
| Excitation/Emission | 340/490, 520 |
| Format | 384-well Plate |
| Product Line | LanthaScreen |
| Reactivity | Human |
Description
- A part of the activating protein-1 (AP-1) transciption factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
- For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/GFP (EmGFP) | |
| 384-well Plate | |
| Human | |
| Dry Ice | |
| TR-FRET |
| 340/490, 520 | |
| LanthaScreen | |
| End Point | |
| Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title