Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody Kit
For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications.
Brand: Thermo Scientific™ PV4454
This item is not returnable.
View return policy
Description
- A part of the activating protein-1 (AP-1) transciption factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
- For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/GFP (EmGFP) | |
| Terbium-labeled anti-c-Jun [pSer73] antibody in HEPES buffered saline. Store at -20°C. | |
| 340/490, 520 | |
| Dry Ice | |
| Hu | |
| TR-FRET | |
| LanthaScreen |
| 1 mg | |
| 384-well Plate | |
| Human | |
| End Point | |
| Serine/Threonine Kinases | |
| MAPK8, MAPK9, MAPK10, NLK |