Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
£726.00 - £8990.00
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10419603
|
Thermo Scientific™
PV4752 |
25 μg |
£726.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10504424
|
Thermo Scientific™
PV4751 |
1 mg |
£8990.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin. This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen™ cell lines to develop robust high-throughput biochemical or cell based assays for ubiquitination or deubiquitination.For Research Use Only. Not for use in diagnostic procedures.