missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
Brand: Thermo Scientific™ PV4752
This item is not returnable.
View return policy
Description
The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin. This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen™ cell lines to develop robust high-throughput biochemical or cell based assays for ubiquitination or deubiquitination.
Specifications
| Biochemical Proteasome Assays | |
| LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit, 25μg, Human Species Reactivity, Tb (Terbium) Conjugate, 340/490nm Absorbance, 520nm Excitation, HEPES buffered saline Storage Buffer, Used for Pharma and Biopharma, Target and Lead Identification | |
| Purified | |
| RUO | |
| Primary | |
| Terbium-labeled anti-ubiquitin-FK2 antibody in HEPES buffered saline. Store at -20°C. | |
| Liquid | |
| 340⁄490, 520 | |
| Human | |
| Hu |
| Tb (Terbium)/Lanthanide Chelates | |
| 150kDa | |
| 25 μg | |
| Pharma and Biopharma | |
| 1 Kit(s) | |
| 150000 Da | |
| 384-well Plate | |
| Microplate Reader | |
| Dry Ice | |
| LanthaScreen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction