Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody Kit
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody Kit
£3740.00 - £18050.00
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10525484
|
Thermo Scientific™
PV5130 |
25 μg |
£3740.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10206794
|
Thermo Scientific™
PV5131 |
1 mg |
£18050.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (PV5132) to evaluate inhibitors in medium to high throughput screening applications.