Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody Kit
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody Kit
Brand: Thermo Scientific™ PV5131
This item is not returnable.
View return policy
Description
- The p53 tumor suppressor protein is an essential component of many different signaling pathways
- The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR
- For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (PV5132) to evaluate inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled anti-phospho-p53 [pSer15] antibody in HEPES buffered saline. Store at -20°C. | |
| 340/490, 520 | |
| Dry Ice | |
| Hu | |
| TR-FRET |
| 1 mg | |
| 384-well Plate | |
| Human | |
| End Point | |
| Serine/Threonine Kinases | |
| LanthaScreen |