missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-LRRK2 [pSer935] Antibody
This assay format provides a convenient tool for the expression of LRRK2-GFP fusion protein in the cell background of the user's choice. The resulting cellular assay can be used to identify LRRK2 and relevant mutant inhibitor.
£4245.00 - £22610.00
Specifications
| For Use With (Equipment) | HTS Reader |
|---|---|
| Host Species | Rabbit |
| Product Line | LanthaScreen |
| Reactivity | Human |
| Shipping Condition | Dry Ice |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
13335554
|
Thermo Scientific™
A14690 |
25 μg |
£4245.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
13345554
|
Thermo Scientific™
A14693 |
100 μg |
£9060.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
13355554
|
Thermo Scientific™
A14694 |
1 mg |
£22610.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The LanthaScreen™ Tb-Anti-LRRK2 [pSer935] Antibody allows the investigation of phosphorylation at serine 935 on LRRK2 protein kinase
- When paired with LRRK2 BacMam reagents, users have the freedom to choose the cellular background for their assay, including primary cells
- This enables screening for potential inhibitors of LRRK2 proteins in their natural complexes in a physiologically relevant cell type
- This antibody has been specifically developed and optimized for use in our LanthaScreen™ TR-FRET LRRK2 Cellular Activity Assay Kits
- The antibody, as well as other components of these assay kits, are also available separately
- Can be used by Parkinson's and other disease researchers to perform HTS, cell-based assays to generate inhibition curves (IC50) for their compound libraries
- This will aid drug discovery efforts by reducing the time to results from 4 hrs to typically 1.5 hrs compared to alternative ELISAs
- For research use only
- Not intended for animal or human diagnostic or therapeutic use
Specifications
| HTS Reader | |
| LanthaScreen | |
| Dry Ice | |
| Phospho |
| Rabbit | |
| Human | |
| Monoclonal |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title