missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody
The LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody is a key component of the BacMam Histone H3K4me3 Cellular Assay (A14163)
£6720.00 - £31670.00
Specifications
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
|---|---|
| Concentration | 3500 nM, 0.5 mg/mL |
| Form | Liquid |
| Shipping Condition | Dry Ice |
| Target Modification State | Methylated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11682049
|
Thermo Scientific™
A14157 |
100 μg |
£6720.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11612059
|
Thermo Scientific™
A14167 |
1 mg |
£31670.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody is a key component of the BacMam Histone H3K4me3 Cellular Assay (A14163)
- The LanthaScreen™ Histone H3K4me3 antibody allows investigation of trimethylation at lysine 4 on histone H3
- This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys4 trimethylation in their natural complexes
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody Lets You:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Liquid | |
| Methylated |
| 3500 nM, 0.5 mg/mL | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title