Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody
The LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody is a key component of the BacMam Histone H3K4me3 Cellular Assay (A14163)
Brand: Thermo Scientific™ A14167
This item is not returnable.
View return policy
Description
- The LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody is a key component of the BacMam Histone H3K4me3 Cellular Assay (A14163)
- The LanthaScreen™ Histone H3K4me3 antibody allows investigation of trimethylation at lysine 4 on histone H3
- This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys4 trimethylation in their natural complexes
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody Lets You:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifications
| 3500 nM, 0.5 mg/mL | |
| 1 mg | |
| Liquid | |
| Methylated |
| Tb (Terbium)/Lanthanide Chelates | |
| Store the LanthaScreen™ Tb-anti-histone H3K4me3 Antibody at -20°C. | |
| Dry Ice |
Product Content Correction
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering