missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-AKT [pSer473] Antibody Kit

LanthaScreen™ Tb-labeled phospho-specific antibodies allow for the detection of intracellular post-translational modifications of specific target proteins.

£2215.00 - £14470.00

Specifications

Conjugate Tb (Terbium)/Lanthanide Chelates
Concentration 0.5 mg/mL
For Use With (Equipment) Microplate Reader
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10083013
View Documents
Thermo Scientific™
PV5267
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10594624
View Documents
Thermo Scientific™
PV5268
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • LanthaScreen™ Tb-labeled phospho-specific antibodies allow for the detection of intracellular post-translational modifications of specific target proteins
  • To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies
  • These antibodies may be accessed in small scale and screening quantities
Specifications

Specifications

Tb (Terbium)/Lanthanide Chelates
Microplate Reader
Rabbit
Dry Ice
0.5 mg/mL
Liquid
Human
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.