missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-AKT [pSer473] Antibody Kit

Product Code. 10083013
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083013 25 μg Each
10594624 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10083013 Supplier Thermo Scientific™ Supplier No. PV5267

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LanthaScreen™ Tb-labeled phospho-specific antibodies allow for the detection of intracellular post-translational modifications of specific target proteins.

  • LanthaScreen™ Tb-labeled phospho-specific antibodies allow for the detection of intracellular post-translational modifications of specific target proteins
  • To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies
  • These antibodies may be accessed in small scale and screening quantities
TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Concentration 0.5 mg/mL
Conjugate Tb (Terbium)/Lanthanide Chelates
Host Species Rabbit
Purification Method Purified
Quantity 25 μg
Content And Storage Terbium labeled anti-AKT [pSer473] antibody in HEPES buffered saline (137 mM NaCl, 2.7 mM KCl, 10 mM HEPES pH 7.5). Storage is at -20°C
Form Liquid
Isotype IgG1
For Use With (Equipment) Microplate Reader
Host abrv Rb
Marker 0.5 mg/mL
Reactivity Human
Shipping Condition Dry Ice
Reactivity abrv Hu
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.