missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ STAT5 TF-1 Cell Line

Product Code. 10583284
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10583284

Brand: Thermo Scientific™ K1598

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LanthaScreen™ GFP Cellular Assays allow for the analysis of post-translational modifications for a number of target proteins in an live-cell context.

LanthaScreen™ GFP Cellular Assays allow for the analysis of post-translational modifications for a number of target proteins in an live-cell context. In hematopoeitic cells, the JAK2/STAT5 signaling pathway plays an essential role in blood cell formation in response to cytokines such as GM-CSF, IL-3, and EPO. In this pathway, binding of these cytokines to their respective cell surface receptors results in the activation of JAK2, which in turn phospho-activates STAT5 proteins at specific tyrosine residues (Tyr-694/699).

LanthaScreen™-STAT5 TF-1 is a human hematopoeitic cell line which constitutively expresses GFP-STAT5 fusion proteins. The JAK2/STAT5 signaling pathway is known to be functionally intact in this cell line, therefore the GFP-STAT5 fusion protein serves as a substrate for the inducible phosphorylation by JAK2. Using this cell line, a homogenous immuno-assay has been developed in which the phosphorylation state of GFP-STAT5 is detected in cell lysates using a LanthaScreen™ Terbium-anti-mouse and anti-phospho STAT5 [pTyr694/699] antibody pair, in a time-resolved FRET (TR-FRET) readout. GFP-STAT5 Lentivirus was transduced into TF-1 cells followed by selection with Blasticidine.

This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as sorting marker, and has been screened for the constitutive expression of GFP-STAT5 fusion protein. Using a lytic TR-FRET immuno-assay, this cell line is validated for EC50 and Z' under optimized conditions using GM-CSF as an agonist for JAK2 -mediated GFP-STAT5 phosphorylation. This assay has also been tested for assay performance under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance and lysis/equilibration time.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists IL-3, EPO
Antagonists JAK Inhibitor
Assay Entry Cell-based terbium phospho-specific activity (stable)
Cell State Dividing Cells
Content And Storage LanthaScreen™ STAT5 TF-1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Description LanthaScreen™ STAT5 TF-1 Cell Line, 1Vial, Fluorescent Detection Method, Kinases, Signaling Pathway Druggable Target, LanthaScreen™ System, End Point Readout, Liquid nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, cellSensor™ cellular pathway assays
Gene Symbol STAT5A
Optimal Response GM-CSF (EC50 = 0.33 ng/mL)
Primary Agonist GM-CSF
Product Line LanthaScreen™
Quantity 1 Vial
Readout TR-FRET
Reporter Gene GFP (EmGFP)
Sample Type TF-1
Target Entry STAT5
Type Cell Line
Cell Line TF-1
System Type LanthaScreen™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.