Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-GST (Rabbit) Antibody
LanthaScreen™ europium-labeled anti-GST rabbit antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling negatively impacts protein activity.
Brand: Thermo Scientific™ A15116
Description
- LanthaScreen™ europium-labeled anti-GST rabbit antibodies are used to indirectly label proteins tagged with GST
- This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity
- These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays
Specifications
| GST | |
| Polyclonal | |
| Rabbit | |
| RUO | |
| Primary | |
| Frozen | |
| Rb | |
| GST | |
| LanthaScreen |
| Kinase Assay | |
| Europium | |
| 100 μg | |
| Drug Development | |
| Europium-labeled anti-GST antibody is supplied in HEPES-buffered saline. Store at -20°C. | |
| Microplate Reader | |
| Drug Development | |
| KA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.