Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-GST (Rabbit) Antibody
LanthaScreen™ europium-labeled anti-GST rabbit antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling negatively impacts protein activity.
Brand:  Thermo Scientific™ A15115
 View more versions of this product
Product Code. 15285684
  • £1022.00 / 25µg
Description
  • LanthaScreen™ europium-labeled anti-GST rabbit antibodies are used to indirectly label proteins tagged with GST
  • This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity
  • These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GST
Polyclonal
Rabbit
RUO
Primary
Frozen
Rb
GST
LanthaScreen
Kinase Assay
Europium
25 μg
Drug Development
Europium-labeled anti-GST antibody is supplied in HEPES-buffered saline. Store at -20°C.
Microplate Reader
Drug Development
KA