Learn More
Invitrogen™ LAMP5 (BAD-LAMP) Monoclonal Antibody (34.2), eBioscience™
Rat Monoclonal Antibody
£111.00 - £1392.00
Specifications
| Antigen | LAMP5 (BAD-LAMP) |
|---|---|
| Clone | 34.2 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17180782
|
Invitrogen™
14-9778-37 |
2 mg |
£1392.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15587676
|
Invitrogen™
14-9778-80 |
25 μg |
£111.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation. This 34.2 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using high or low pH antigen retrieval and can be used at less than or equal to 1 μg/mL. This 34.2 antibody has been tested by western blot analysis of reduced SK-N-SH cell lysates and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Lysosome-associated membrane protein 5 (LAMP5) is a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.Specifications
| LAMP5 (BAD-LAMP) | |
| 0.5 mg/mL | |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.09% Sodium Azide; pH 7.2 | |
| Q9UJQ1 | |
| 24141 | |
| Primary | |
| Affinity chromatography |
| 34.2 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| LAMP5 | |
| 3110035N03Rik; 6330527O06Rik; BADLAMP; BAD-LAMP; brain and dendritic cell associated LAMP; brain and dendritic cell-associated LAMP; Brain-associated LAMP-like protein; C20orf103; LAMP family protein C20orf103; LAMP family protein C20orf103 homolog; LAMP5; LAMP-5; lysosomal associated membrane protein family member 5; lysosomal-associated membrane protein family, member 5; lysosome-associated membrane glycoprotein 5; Lysosome-associated membrane protein 5; Protein C20orf103; RGD1306991; UNC-46 | |
| LAMP5 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.