missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LAMP5 (BAD-LAMP) Monoclonal Antibody (34.2), eBioscience™

Rat Monoclonal Antibody

Brand:  Invitrogen™ 14-9778-80

 View more versions of this product

Product Code. 15587676

  • £111.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation. This 34.2 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using high or low pH antigen retrieval and can be used at less than or equal to 1 μg/mL. This 34.2 antibody has been tested by western blot analysis of reduced SK-N-SH cell lysates and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Lysosome-associated membrane protein 5 (LAMP5) is a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAMP5 (BAD-LAMP)
Monoclonal
0.5 mg/mL
PBS with 0.09% Sodium Azide; pH 7.2
Q9UJQ1
LAMP5
Affinity chromatography
RUO
24141
4°C
Liquid
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
34.2
Unconjugated
LAMP5
3110035N03Rik; 6330527O06Rik; BADLAMP; BAD-LAMP; brain and dendritic cell associated LAMP; brain and dendritic cell-associated LAMP; Brain-associated LAMP-like protein; C20orf103; LAMP family protein C20orf103; LAMP family protein C20orf103 homolog; LAMP5; LAMP-5; lysosomal associated membrane protein family member 5; lysosomal-associated membrane protein family, member 5; lysosome-associated membrane glycoprotein 5; Lysosome-associated membrane protein 5; Protein C20orf103; RGD1306991; UNC-46
Rat
25 μg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.