missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£317.00 - £473.00
Specifications
| Antigen | Laminin beta 3 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630795
|
Novus Biologicals
NBP3-21221-25ul |
25 μL |
£317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659883
|
Novus Biologicals
NBP3-21221-100ul |
100 μL |
£473.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Laminin beta 3 Polyclonal antibody specifically detects Laminin beta 3 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Laminin beta 3 | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KLVTSMTKQLGDFWTRMEELRHQARQQGAEAVQAQQLAEGASEQALSAQEGFERIKQKYAELKDRLGQSSMLGEQGARIQSVKTEAEELFGETMEMMDRMKDMELELLRGSQAIMLRSADLTGLEKRVEQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 3914 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title