missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21221-25ul
This item is not returnable.
View return policy
Description
Laminin beta 3 Polyclonal antibody specifically detects Laminin beta 3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Laminin beta 3 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KLVTSMTKQLGDFWTRMEELRHQARQQGAEAVQAQQLAEGASEQALSAQEGFERIKQKYAELKDRLGQSSMLGEQGARIQSVKTEAEELFGETMEMMDRMKDMELELLRGSQAIMLRSADLTGLEKRVEQ | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3914 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction