missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Laminin alpha 3/Laminin-5 Antibody (SR3297), Novus Biologicals™

Rabbit Monoclonal Antibody

£240.00 - £290.00

Specifications

Antigen Laminin alpha 3/Laminin-5
Clone SR3297
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30041732
View Documents
Novus Biologicals™
NBP3-27766-50ul
50 μL
£240.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30041523
View Documents
Novus Biologicals™
NBP3-27766-100ul
100 μL
£290.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Laminin alpha 3/Laminin-5 Monoclonal antibody specifically detects Laminin alpha 3/Laminin-5 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Laminin alpha 3/Laminin-5
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
PBS (pH 7.4), 150mM NaCl, 50% glycerol
A synthesized peptide derived from human Laminin alpha 3/Laminin-5 (Uniprot #: Q16787)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
SR3297
Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Human
a]3B;BM600;E170;LAMA3;Lama3B;LAMNA;LOCS
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.