missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Laminin alpha 3/Laminin-5 Antibody (SR3297), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27766-100ul
This item is not returnable.
View return policy
Description
Laminin alpha 3/Laminin-5 Monoclonal antibody specifically detects Laminin alpha 3/Laminin-5 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Laminin alpha 3/Laminin-5 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3297 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| a]3B;BM600;E170;LAMA3;Lama3B;LAMNA;LOCS | |
| A synthesized peptide derived from human Laminin alpha 3/Laminin-5 (Uniprot #: Q16787) | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction