missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 2 Antibody (CL3450), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46624-25ul
This item is not returnable.
View return policy
Description
Laminin alpha 2 Monoclonal antibody specifically detects Laminin alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Laminin alpha 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMMLaminin-4 subunit alpha, Merosin heavy chain | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence DKLKPIKELEDNLKKNISEIKELINQARKQANSIKVSVSSGGDCIRTYKPEIKKGSYNNIVVNVKTAVADNLLFYLGSAKFIDFLAIEMRKGKVSFLWDVGSGVGRVEYPDLTIDDSYWYRIVASRTGRNGT | |
| 25 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3450 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| P24043 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3908 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction