missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B1 Antibody (919022), Unconjugated - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£194.00 - £403.00
Specifications
| Antigen | Lamin B1 |
|---|---|
| Clone | 919022 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1 - 2 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602144
|
Novus Biologicals
MAB85251 |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682914
|
Novus Biologicals
MAB85251-SP |
25 μg |
£194.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lamin B1 Monoclonal antibody specifically detects Lamin B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Lamin B1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors | |
| PBS | |
| 4001 | |
| IgG2b | |
| Protein A or G purified |
| 919022 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1 - 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419 | |
| E. coli-derived partial recombinant human Lamin B1 protein (amino acids 471-582) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title