missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B1 Antibody (919022), Unconjugated - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB85251-SP
This item is not returnable.
View return policy
Description
Lamin B1 Monoclonal antibody specifically detects Lamin B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Lamin B1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1 - 2 μg/mL | |
| ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419 | |
| E. coli-derived partial recombinant human Lamin B1 protein (amino acids 471-582) | |
| 25 μg | |
| Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors | |
| 4001 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 919022 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction