Learn More
Lamin B1 Antibody (919022), Unconjugated - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals MAB85251
 View more versions of this product
Product Code. 18602144
  • £403.00 / 100µg
Description
Lamin B1 Monoclonal antibody specifically detects Lamin B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lamin B1
Monoclonal
1.0 mg/mL
Western Blot 1:100 - 1:2000, Immunohistochemistry 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1 - 2 μg/mL
ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419
E. coli-derived partial recombinant human Lamin B1 protein (amino acids 471-582)
100 μg
Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors
4001
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
919022
Unconjugated
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified