missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lamin B Receptor Recombinant Rabbit Monoclonal Antibody (23GB6100), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Lamin B Receptor |
|---|---|
| Clone | 23GB6100 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087643
![]() |
Invitrogen™
MA552984 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087623
|
Invitrogen™
MA552985 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lamin B Receptor Recombinant Monoclonal antibody specifically detects Lamin B Receptor in Human, Rat samples. It is validated for Western BlotSpecifications
| Lamin B Receptor | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| O08984, Q14739 | |
| 3930, 89789 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6100 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 3-beta-hydroxysterol Delta (14)-reductase; AI505894; C-14 sterol reductase; C14SR; Delta(14)-sterol reductase LBR; delta(14)-sterol reductase LBR; lamin-B receptor; Delta-14-SR; DHCR14B; FLJ43126; ic; ichthyosis; integral nuclear envelope inner membrane protein; lamin B receptor; lamin-B receptor; lamin-b receptor-like protein; Lbr; LMN2R; MGC9041; NBP60; PHA; PRO0650; sb:cb406; Sterol C14-reductase; TDRD18; tudor domain containing 18; wu:fb75h08; wu:fc47b04; wu:fd36b07; zgc:86649 | |
| A synthesized peptide derived from human Lamin B Receptor (300-615AA). | |
| Primary | |
| -20°C | |
| LBR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title