missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lamin B Receptor Recombinant Rabbit Monoclonal Antibody (23GB6100), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552984
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
| Lamin B Receptor | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LBR | |
| 3-beta-hydroxysterol Delta (14)-reductase; AI505894; C-14 sterol reductase; C14SR; Delta(14)-sterol reductase LBR; delta(14)-sterol reductase LBR; lamin-B receptor; Delta-14-SR; DHCR14B; FLJ43126; ic; ichthyosis; integral nuclear envelope inner membrane protein; lamin B receptor; lamin-B receptor; lamin-b receptor-like protein; Lbr; LMN2R; MGC9041; NBP60; PHA; PRO0650; sb:cb406; Sterol C14-reductase; TDRD18; tudor domain containing 18; wu:fb75h08; wu:fc47b04; wu:fd36b07; zgc:86649 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3930, 89789 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6100 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08984, Q14739 | |
| LBR | |
| A synthesized peptide derived from human Lamin B Receptor (300-615AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction