Learn More
Lamin A/C Rabbit, anti-Human, CoraLite Plus 488, Clone: 5I16, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | Lamin A/C |
|---|---|
| Clone | 5I16 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084295
|
Proteintech
CL488-81042-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18077075
|
Proteintech
CL488-81042-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Lamin A/C antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Lamin A/C. This antibody recognizes human, mouse, rat antigen. The Lamin A/C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Lamin A/C | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 4000 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 5I16 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| 70 kDa lamin, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, lamin A, lamin A/C, LDP1, LFP, LGMD1B, LMN1, LMNA, LMNC, Prelamin A/C, PRO1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.