Learn More
Lamin A/C Rabbit, anti-Human, CoraLite Plus 488, Clone: 5I16, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-81042-100UL
Description
The Lamin A/C antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Lamin A/C. This antibody recognizes human, mouse, rat antigen. The Lamin A/C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Lamin A/C | |
| 5I16 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| 70 kDa lamin, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, lamin A, lamin A/C, LDP1, LFP, LGMD1B, LMN1, LMNA, LMNC, Prelamin A/C, PRO1 | |
| 100 μL | |
| 4000 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |