missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRP19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98571-50ul
This item is not returnable.
View return policy
Description
PRP19 Polyclonal antibody specifically detects PRP19 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| PRP19 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:50-1:200 | |
| hPSO4, NMP200SNEV, Nuclear matrix protein 200, nuclear matrix protein NMP200 related to splicing factor PRP19, pre-mRNA-processing factor 19, PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae), PRP19PRP19/PSO4 homolog (S. cerevisiae), PSO4PRP19/PSO4 homolog, Senescence evasion factor, UBOX4 | |
| Produced in rabbits immunized with E. coli-derived Human PRP19 fragment. | |
| 50 μg | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| 27339 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction