missing translation for 'onlineSavingsMsg'
Learn More

PRP19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen PRP19
Dilution Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:50-1:200
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652462
View Documents
Novus Biologicals
NBP2-98571-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18600485
View Documents
Novus Biologicals
NBP2-98571-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRP19 Polyclonal antibody specifically detects PRP19 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

PRP19
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Cancer, DNA Repair, Tumor Suppressors
PBS (pH 7.0)
27339
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
Human
hPSO4, NMP200SNEV, Nuclear matrix protein 200, nuclear matrix protein NMP200 related to splicing factor PRP19, pre-mRNA-processing factor 19, PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae), PRP19PRP19/PSO4 homolog (S. cerevisiae), PSO4PRP19/PSO4 homolog, Senescence evasion factor, UBOX4
Produced in rabbits immunized with E. coli-derived Human PRP19 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.